Assay Details
Target Gene Details
Entrez Gene ID: | 9140 |
Gene Name: | autophagy related 12 |
Gene Aliases: |
APG12, APG12L, FBR93, HAPG12 |
Location: |
Chr.5:115828196-115841851 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG12 | NM_001277783.1 | NP_001264712.1 | ||
| NM_004707.3 | NP_004698.3 | |||
| NR_033362.1 | ||||
| NR_033363.1 | ||||
| NR_073603.1 | ||||
| AB017507.1 | BAA36493.1 | |||
| AK313320.1 | ||||
| BC011033.1 | AAH11033.1 | |||
| BC012266.2 | AAH12266.2 | |||
| BP335591.1 | ||||
| CB995198.1 | ||||
| CN400030.1 | ||||
| CR978239.1 | ||||
| DA005347.1 | ||||
| DB048378.1 | ||||
| EU554057.1 |
More Information
Set Membership: |
|

Genomic Map