Assay Details
Target Gene Details
Entrez Gene ID: | 2561 |
Gene Name: | gamma-aminobutyric acid type A receptor beta2 subunit |
Gene Aliases: |
- |
Location: |
Chr.5:161288429-161548124 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABRB2 | NM_000813.2 | NP_000804.1 | ||
| NM_021911.2 | NP_068711.1 | |||
| A28108.1 | CAA01922.1 | |||
| AK289730.1 | ||||
| AK289815.1 | ||||
| AK294437.1 | ||||
| BC099705.1 | AAH99705.1 | |||
| BC099719.1 | AAH99719.1 | |||
| BC105639.1 | AAI05640.1 | |||
| GU086163.1 | ||||
| S67368.1 | AAB29370.1 | |||
| S77553.1 | AAB33982.1 | |||
| S77554.1 | AAB33983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5100 | Chr.5:161264110 - 161308790 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113347700] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map