Assay Details
Target Gene Details
Entrez Gene ID: | 27039 |
Gene Name: | polycystin 2 like 2, transient receptor potential cation channel |
Gene Aliases: |
TRPP5 |
Location: |
Chr.5:137889420-137942747 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKD2L2 | NM_001258448.1 | NP_001245377.1 | ||
| NM_001258449.1 | NP_001245378.1 | |||
| NM_001300921.1 | NP_001287850.1 | |||
| NM_014386.3 | NP_055201.2 | |||
| XM_011543318.2 | XP_011541620.1 | |||
| XM_011543321.1 | XP_011541623.1 | |||
| XM_017009343.1 | XP_016864832.1 | |||
| XM_017009344.1 | XP_016864833.1 | |||
| XM_017009345.1 | XP_016864834.1 | |||
| AF118125.1 | AAD46478.1 | |||
| AF182034.1 | AAF65622.1 | |||
| AK301924.1 | ||||
| BC044581.1 | AAH44581.1 | |||
| BG719682.1 | ||||
| BG721989.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830501 | Chr.5:137788777 - 137977035 on Build GRCh38 | Gain |
|
| esv3606913 | Chr.5:137856857 - 138124533 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76734697] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map