Assay Details
Target Gene Details
Entrez Gene ID: | 140890 |
Gene Name: | splicing regulatory glutamic acid and lysine rich protein 1 |
Gene Aliases: |
SFRS12, SRrp508, SRrp86 |
Location: |
Chr.5:66144218-66183616 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SREK1 | NM_001077199.2 | NP_001070667.1 | ||
| NM_001270492.1 | NP_001257421.1 | |||
| NM_001323527.1 | NP_001310456.1 | |||
| NM_001323529.1 | NP_001310458.1 | |||
| NM_001323533.1 | NP_001310462.1 | |||
| NM_001323534.1 | NP_001310463.1 | |||
| NM_001323535.1 | NP_001310464.1 | |||
| NM_139168.3 | NP_631907.1 | |||
| XM_011543171.2 | XP_011541473.1 | |||
| XM_017009055.1 | XP_016864544.1 | |||
| XM_017009056.1 | XP_016864545.1 | |||
| AF459094.1 | AAL67778.1 | |||
| AK091758.1 | ||||
| AK308287.1 | ||||
| AL832888.1 | CAH10414.1 | |||
| BC047322.1 | AAH47322.1 | |||
| BC067770.1 | ||||
| BC112343.1 | AAI12344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830328 | Chr.5:66088707 - 66295526 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map