Assay Details
Target Gene Details
Entrez Gene ID: | 84868 |
Gene Name: | hepatitis A virus cellular receptor 2 |
Gene Aliases: |
CD366, HAVcr-2, KIM-3, TIM3, TIMD-3, TIMD3, Tim-3 |
Location: |
Chr.5:157085832-157109237 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAVCR2 | NM_032782.4 | NP_116171.3 | ||
| AB924452.1 | ||||
| AF251707.1 | AAM19100.1 | |||
| AF450242.1 | AAL65157.1 | |||
| AF450243.1 | AAL65158.1 | |||
| AK027334.1 | BAB55044.1 | |||
| AK130144.1 | ||||
| AK314406.1 | ||||
| AY069944.1 | AAL55401.1 | |||
| BC063431.1 | AAH63431.1 | |||
| JX049979.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575892 | Chr.5:156975204 - 157220778 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map