Assay Details
Target Gene Details
Entrez Gene ID: | 9337 |
Gene Name: | CCR4-NOT transcription complex subunit 8 |
Gene Aliases: |
CAF1, CALIF, Caf1b, POP2, hCAF1 |
Location: |
Chr.5:154857952-154876792 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
More Information
Additional Information:
For this assay, SNP(s) [rs114494038] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map