Assay Details
Target Gene Details
Entrez Gene ID: | 7322 |
Gene Name: | ubiquitin conjugating enzyme E2 D2 |
Gene Aliases: |
E2(17)KB2, PUBC1, UBC4, UBC4/5, UBCH4, UBCH5B |
Location: |
Chr.5:139561166-139628434 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2D2 | NM_003339.2 | NP_003330.1 | ||
| NM_181838.1 | NP_862821.1 | |||
| XM_017009820.1 | XP_016865309.1 | |||
| AK001311.1 | ||||
| AK001428.1 | ||||
| AK310273.1 | ||||
| AY651263.1 | AAX35690.1 | |||
| BC033349.1 | AAH33349.1 | |||
| BQ186099.1 | ||||
| U39317.1 | AAA91460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830505 | Chr.5:139551597 - 139745300 on Build GRCh38 | Loss |
|
| nsv519808 | Chr.5:139396890 - 139805924 on Build GRCh38 | Loss |
|
| nsv462460 | Chr.5:139553566 - 139765464 on Build GRCh38 | Loss |
|
| nsv830504 | Chr.5:139511905 - 139701120 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map