Assay Details
Target Gene Details
Entrez Gene ID: | 101927761 |
Gene Name: | T helper type 2 locus control region associated RNA |
Gene Aliases: |
TH2-LCR |
Location: |
Chr.5:132630589-132656154 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TH2LCRR | NR_132125.1 | |||
| NR_132126.1 | 1 | 59 | ||
| AA470013.1 | 1 | 59 | ||
| BX095327.1 | 1 | 58 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950392 | Chr.5:132655609 - 132670708 on Build GRCh38 | Deletion |
|
| nsv528281 | Chr.5:132650771 - 132682252 on Build GRCh38 | Gain |
|

Genomic Map