Assay Details
Target Gene Details
Entrez Gene ID: | 133746 |
Gene Name: | junction mediating and regulatory protein, p53 cofactor |
Gene Aliases: |
WHAMM2, WHDC1L3 |
Location: |
Chr.5:79236102-79327215 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JMY | NM_152405.4 | 1 | 321 | NP_689618.4 |
| XM_005248430.2 | 1 | 321 | XP_005248487.1 | |
| XM_011543155.2 | 1 | 321 | XP_011541457.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762527 | Chr.5:78625376 - 79252373 on Build GRCh38 | Gain |
|
| nsv1019620 | Chr.5:79133218 - 79361144 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map