Assay Details
Target Gene Details
Entrez Gene ID: | 10409 |
Gene Name: | brain abundant membrane attached signal protein 1 |
Gene Aliases: |
CAP-23, CAP23, NAP-22, NAP22 |
Location: |
Chr.5:17216823-17276845 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BASP1 | NM_001271606.1 | NP_001258535.1 | ||
| NM_006317.4 | NP_006308.3 | |||
| AF039656.1 | AAC67374.1 | |||
| AK295995.1 | ||||
| AK314152.1 | ||||
| BC000518.2 | AAH00518.1 | |||
| BG704783.1 | ||||
| CN355202.1 | ||||
| DW458407.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv597312 | Chr.5:17214792 - 17225224 on Build GRCh38 | Loss |
|
| nsv830218 | Chr.5:17093053 - 17289936 on Build GRCh38 | Loss |
|
| nsv823015 | Chr.5:17215900 - 17219090 on Build GRCh38 | Gain |
|
| nsv521697 | Chr.5:17211540 - 17225224 on Build GRCh38 | Loss |
|
| nsv4738 | Chr.5:17178158 - 17223568 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map