Assay Details
Target Gene Details
Entrez Gene ID: | 26762 |
Gene Name: | hepatitis A virus cellular receptor 1 |
Gene Aliases: |
CD365, HAVCR, HAVCR-1, KIM-1, KIM1, TIM, TIM-1, TIM1, TIMD-1, TIMD1 |
Location: |
Chr.5:156979443-157059119 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAVCR1 | XM_017009340.1 | XP_016864829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1009526 | Chr.5:156993570 - 157000957 on Build GRCh38 | Deletion |
|
| esv3607302 | Chr.5:156839545 - 157055054 on Build GRCh38 | Gain |
|
| dgv1661e212 | Chr.5:156994435 - 156999652 on Build GRCh38 | Loss |
|
| esv3607306 | Chr.5:156969763 - 157052820 on Build GRCh38 | Gain |
|
| esv1004108 | Chr.5:156994448 - 156998621 on Build GRCh38 | Loss |
|
| esv3607307 | Chr.5:156994838 - 157000033 on Build GRCh38 | Loss |
|
| esv3575892 | Chr.5:156975204 - 157220778 on Build GRCh38 | Gain |
|
| esv2667486 | Chr.5:156994839 - 156999948 on Build GRCh38 | Deletion |
|
| esv3566863 | Chr.5:156994806 - 157000186 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116364340] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map