Assay Details
Target Gene Details
Entrez Gene ID: | 9542 |
Gene Name: | neuregulin 2 |
Gene Aliases: |
DON1, HRG2, NTAK |
Location: |
Chr.5:139846779-140043675 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRG2 | NM_001184935.1 | NP_001171864.1 | ||
| NM_004883.2 | NP_004874.1 | |||
| NM_013981.3 | NP_053584.1 | |||
| NM_013982.2 | NP_053585.1 | |||
| NM_013983.2 | NP_053586.1 | |||
| XM_005268533.3 | XP_005268590.1 | |||
| XM_006714810.3 | XP_006714873.1 | |||
| XM_017010064.1 | XP_016865553.1 | |||
| XM_017010065.1 | XP_016865554.1 | |||
| XM_017010066.1 | XP_016865555.1 | |||
| AB005060.1 | BAA23417.1 | |||
| AK124504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471051 | Chr.5:139805924 - 139878018 on Build GRCh38 | Gain |
|
| nsv830506 | Chr.5:139837740 - 140020316 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77865538] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map