Assay Details
Target Gene Details
Entrez Gene ID: | 9597 |
Gene Name: | SMAD5 antisense RNA 1 |
Gene Aliases: |
DAMS, SMAD5AS, SMAD5OS |
Location: |
Chr.5:136129513-136134890 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD5-AS1 | NR_026763.1 | |||
| AF071111.1 | AAD20804.1 | |||
| AF086556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759380 | Chr.5:135829912 - 136139310 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs199706251] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map