Assay Details
Target Gene Details
Entrez Gene ID: | 1176 |
Gene Name: | adaptor related protein complex 3 sigma 1 subunit |
Gene Aliases: |
CLAPS3, Sigma3A |
Location: |
Chr.5:115841606-115914081 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP3S1 | NM_001002924.2 | NP_001002924.2 | ||
| NM_001284.3 | NP_001275.1 | |||
| NM_001318090.1 | NP_001305019.1 | |||
| NM_001318091.1 | NP_001305020.1 | |||
| NM_001318093.1 | NP_001305022.1 | |||
| NM_001318094.1 | NP_001305023.1 | |||
| XM_011543136.2 | XP_011541438.2 | |||
| XM_017009024.1 | XP_016864513.1 | |||
| XM_017009025.1 | XP_016864514.1 | |||
| AK311843.1 | ||||
| BC000804.1 | AAH00804.1 | |||
| BI600357.1 | ||||
| BI757703.1 | ||||
| BT009833.1 | AAP88835.1 | |||
| BX394598.2 | ||||
| BX647735.1 | ||||
| CN411347.1 | ||||
| CR450341.1 | CAG29337.1 | |||
| D63643.1 | BAA09798.1 | |||
| DB483797.1 | ||||
| U91932.1 | AAD03779.1 | |||
| X99458.1 | CAA67823.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2730625 | Chr.5:115842079 - 115870022 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map