Assay Details
Target Gene Details
Entrez Gene ID: | 56990 |
Gene Name: | CDC42 small effector 2 |
Gene Aliases: |
SPEC2 |
Location: |
Chr.5:131264009-131394690 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42SE2 | NM_001038702.1 | NP_001033791.1 | ||
| NM_020240.2 | NP_064625.1 | |||
| XM_017009648.1 | XP_016865137.1 | |||
| XM_017009649.1 | XP_016865138.1 | |||
| AK312406.1 | ||||
| BI460220.1 | ||||
| BI522591.1 | ||||
| DA321488.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575881 | Chr.5:130637365 - 131735693 on Build GRCh38 | Gain |
|
| nsv524005 | Chr.5:131262625 - 131324702 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map