Assay Details
Target Gene Details
Entrez Gene ID: | 51307 |
Gene Name: | family with sequence similarity 53 member C |
Gene Aliases: |
C5orf6 |
Location: |
Chr.5:138337535-138349729 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM53C | NM_001135647.1 | NP_001129119.1 | ||
| NM_016605.2 | NP_057689.1 | |||
| AF251040.1 | AAF63766.1 | |||
| AK026729.1 | ||||
| AK027412.1 | ||||
| AK310378.1 | ||||
| AK315567.1 | ||||
| BC000259.1 | AAH00259.1 | |||
| BC052993.1 | AAH52993.1 | |||
| DA216192.1 | ||||
| DB035183.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033604 | Chr.5:138291351 - 138404936 on Build GRCh38 | Gain |
|
| esv3606920 | Chr.5:138339900 - 138344394 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map