Assay Details
Target Gene Details
Entrez Gene ID: | 23305 |
Gene Name: | acyl-CoA synthetase long-chain family member 6 |
Gene Aliases: |
ACS2, FACL6, LACS 6, LACS2, LACS5 |
Location: |
Chr.5:131949973-132012068 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSL6 | NM_001009185.2 | 21 | 6185 | NP_001009185.1 |
| NM_001205247.1 | 21 | 6080 | NP_001192176.1 | |
| NM_001205248.1 | 21 | 6172 | NP_001192177.1 | |
| NM_001205250.1 | 21 | 6182 | NP_001192179.1 | |
| NM_001205251.1 | 20 | 5947 | NP_001192180.1 | |
| NM_015256.3 | 21 | 6185 | NP_056071.2 | |
| BM992189.1 | 1 | 370 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1031854 | Chr.5:131900850 - 131969174 on Build GRCh38 | Gain |
|
| nsv1024604 | Chr.5:131861051 - 132088663 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map