Assay Details
Target Gene Details
Entrez Gene ID: | 101929524 |
Gene Name: | uncharacterized LOC101929524 |
Gene Aliases: |
- |
Location: |
Chr.5:16615926-16629969 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929524 | NR_109946.1 | 4 | 1615 | |
| AK097784.1 | 4 | 1615 |

Genomic Map