Assay Details
Target Gene Details
Entrez Gene ID: | 1729 |
Gene Name: | diaphanous related formin 1 |
Gene Aliases: |
DFNA1, DIA1, DRF1, LFHL1, SCBMS, hDIA1 |
Location: |
Chr.5:141515021-141619055 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIAPH1 | NM_001079812.2 | NP_001073280.1 | ||
| NM_001314007.1 | NP_001300936.1 | |||
| NM_005219.4 | NP_005210.3 | |||
| XM_011537572.2 | XP_011535874.1 | |||
| XM_011537573.2 | XP_011535875.1 | |||
| AB209482.1 | BAD92719.1 | |||
| AF051782.1 | AAC05373.1 | |||
| AK023345.1 | BAB14533.1 | |||
| AK301195.1 | ||||
| AK307822.1 | ||||
| AY363395.1 | AAQ63049.1 | |||
| BC007411.2 | AAH07411.2 | |||
| BC117257.1 | ||||
| BC143413.1 | ||||
| DQ067452.1 | ||||
| DQ067453.1 |
Target Gene Details
Entrez Gene ID: | 100505658 |
Gene Name: | uncharacterized LOC100505658 |
Gene Aliases: |
- |
Location: |
Chr.5:141558311-141565260 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100505658 | NR_038333.1 | 3 | 1204 |
More Information
Set Membership: |
|

Genomic Map