Assay Details
Target Gene Details
Entrez Gene ID: | 115827 |
Gene Name: | RAB3C, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.5:58582188-58859394 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB3C | NM_001317915.1 | NP_001304844.1 | ||
| NM_138453.3 | NP_612462.1 | |||
| AY026936.1 | AAK08968.1 | |||
| BC013033.2 | AAH13033.1 | |||
| BX375794.2 | ||||
| BX394661.2 | ||||
| DB485923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3570158 | Chr.5:58591168 - 58595891 on Build GRCh38 | Loss |
|
| esv3575847 | Chr.5:58547097 - 58605566 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116002153,rs77031569] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map