Assay Details
Target Gene Details
Entrez Gene ID: | 64097 |
Gene Name: | erythrocyte membrane protein band 4.1 like 4A |
Gene Aliases: |
EPB41L4, NBL4 |
Location: |
Chr.5:112141829-112419490 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPB41L4A | XM_011543530.1 | XP_011541832.1 | ||
| XM_011543531.2 | XP_011541833.1 | |||
| XM_011543532.1 | XP_011541834.1 | |||
| XM_011543533.1 | XP_011541835.1 | |||
| AK096057.1 | BAC04690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599367 | Chr.5:111588583 - 112267016 on Build GRCh38 | Gain |
|
| esv3606287 | Chr.5:112058821 - 112193605 on Build GRCh38 | Gain |
|
| dgv1124e214 | Chr.5:112057248 - 112193605 on Build GRCh38 | Loss |
|
| nsv599368 | Chr.5:112064188 - 112696978 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73227498] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map