Assay Details
Target Gene Details
Entrez Gene ID: | 85021 |
Gene Name: | RALBP1 associated Eps domain containing 1 |
Gene Aliases: |
RALBP1 |
Location: |
Chr.6:138904015-138988282 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REPS1 | NM_001128617.2 | NP_001122089.1 | ||
| NM_001286611.1 | NP_001273540.1 | |||
| NM_001286612.1 | NP_001273541.1 | |||
| NM_031922.4 | NP_114128.3 | |||
| XM_005267177.3 | XP_005267234.1 | |||
| XM_005267178.4 | XP_005267235.1 | |||
| XM_005267179.3 | XP_005267236.1 | |||
| XM_011536202.2 | XP_011534504.1 | |||
| XM_017011387.1 | XP_016866876.1 | |||
| XM_017011388.1 | XP_016866877.1 | |||
| XM_017011389.1 | XP_016866878.1 | |||
| AA706721.1 | ||||
| AB209143.1 | BAD92380.1 | |||
| AF251052.1 | AAK34942.1 | |||
| AJ420514.1 | ||||
| AK312938.1 | ||||
| AL831900.1 | CAD38569.1 | |||
| AL832307.1 | ||||
| BC012764.2 | AAH12764.1 | |||
| BC021211.2 | AAH21211.1 |
More Information
Set Membership: |
|

Genomic Map