Assay Details
Target Gene Details
Entrez Gene ID: | 100130476 |
Gene Name: | uncharacterized LOC100130476 |
Gene Aliases: |
- |
Location: |
Chr.6:137823669-137868233 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130476 | NR_049793.1 | |||
| AK124173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830818 | Chr.6:137790698 - 137950293 on Build GRCh38 | Gain |
|
| nsv830819 | Chr.6:137841569 - 138013923 on Build GRCh38 | Loss |
|
| nsv470863 | Chr.6:137080640 - 138014159 on Build GRCh38 | Loss |
|

Genomic Map