Assay Details
Target Gene Details
Entrez Gene ID: | 134829 |
Gene Name: | clavesin 2 |
Gene Aliases: |
C6orf212, C6orf213, RLBP1L2, bA160A10.4 |
Location: |
Chr.6:122993693-123081282 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLVS2 | XM_017010266.1 | 1 | 1842 | XP_016865755.1 |
More Information
Set Membership: |
|

Genomic Map