Assay Details
Target Gene Details
Entrez Gene ID: | 7280 |
Gene Name: | tubulin beta 2A class IIa |
Gene Aliases: |
CDCBM5, TUBB, TUBB2 |
Location: |
Chr.6:3153666-3157549 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBB2A | NM_001069.2 | NP_001060.1 | ||
| NM_001310315.1 | NP_001297244.1 | |||
| AK312621.1 | ||||
| AY159127.1 | AAN85571.1 | |||
| BC001194.1 | AAH01194.1 | |||
| BC018780.1 | AAH18780.1 | |||
| BC141924.1 | ||||
| BC141925.1 | ||||
| BM846690.1 | ||||
| BP381259.1 | ||||
| CR541958.1 | CAG46756.1 | |||
| X79535.1 | CAA56071.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462614 | Chr.6:3152171 - 3204587 on Build GRCh38 | Gain |
|
| nsv965675 | Chr.6:3154696 - 3158027 on Build GRCh38 | Duplication |
|
| esv2731488 | Chr.6:3155434 - 3180070 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116267518] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map