Assay Details
Target Gene Details
Entrez Gene ID: | 135112 |
Gene Name: | nuclear receptor coactivator 7 |
Gene Aliases: |
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3 |
Location: |
Chr.6:125780948-125932030 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCOA7 | NM_001122842.2 | NP_001116314.1 | ||
| NM_001199619.1 | NP_001186548.1 | |||
| NM_001199620.1 | NP_001186549.1 | |||
| NM_001199621.1 | NP_001186550.1 | |||
| NM_181782.4 | NP_861447.3 | |||
| XM_005266822.4 | XP_005266879.1 | |||
| XM_006715340.3 | XP_006715403.1 | |||
| XM_011535455.2 | XP_011533757.1 | |||
| XM_017010270.1 | XP_016865759.1 | |||
| XM_017010271.1 | XP_016865760.1 | |||
| XM_017010273.1 | XP_016865762.1 | |||
| XM_017010274.1 | XP_016865763.1 | |||
| AB074156.1 | BAE45731.1 | |||
| AB074157.1 | BAE45732.1 | |||
| AF493978.1 | AAM27392.1 | |||
| AK127512.1 | ||||
| AK294558.1 | ||||
| AK307395.1 | ||||
| AL832621.2 | CAD90002.2 | |||
| AL832628.2 | CAD89948.1 | |||
| BC036461.1 | AAH36461.1 | |||
| BC137094.1 | ||||
| BC137095.1 | ||||
| BX537385.1 | CAD97627.1 | |||
| DA770225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950505 | Chr.6:125772255 - 125803354 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map