Assay Details
Target Gene Details
Entrez Gene ID: | 3720 |
Gene Name: | jumonji and AT-rich interaction domain containing 2 |
Gene Aliases: |
JMJ |
Location: |
Chr.6:15245975-15522042 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JARID2 | NM_001267040.1 | NP_001253969.1 | ||
| NM_004973.3 | NP_004964.2 | |||
| XM_005249089.3 | XP_005249146.1 | |||
| XM_011514579.2 | XP_011512881.1 | |||
| XM_017010833.1 | XP_016866322.1 | |||
| XM_017010834.1 | XP_016866323.1 | |||
| AK292861.1 | ||||
| AK302624.1 | ||||
| AK303610.1 | ||||
| BC046246.1 | AAH46246.1 | |||
| U57592.1 | AAC50822.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830594 | Chr.6:15170927 - 15261065 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79777194] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map