Assay Details
Target Gene Details
Entrez Gene ID: | 135154 |
Gene Name: | succinate dehydrogenase complex assembly factor 4 |
Gene Aliases: |
C6orf57, Sdh8 |
Location: |
Chr.6:70566922-70588903 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDHAF4 | NM_145267.2 | NP_660310.2 | ||
| BC018085.1 | AAH18085.1 | |||
| BC104649.1 | AAI04650.1 | |||
| BG703280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524328 | Chr.6:70252130 - 70849102 on Build GRCh38 | Gain |
|
| nsv1027049 | Chr.6:70251907 - 70851785 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map