Assay Details
Target Gene Details
Entrez Gene ID: | 5961 |
Gene Name: | peripherin 2 |
Gene Aliases: |
AOFMD, AVMD, CACD2, DS, MDBS1, PRPH, RDS, RP7, TSPAN22, rd2 |
Location: |
Chr.6:42694509-42723024 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPH2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830650 | Chr.6:42690493 - 42873400 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|

Genomic Map