Assay Details
Target Gene Details
Entrez Gene ID: | 58528 |
Gene Name: | Ras related GTP binding D |
Gene Aliases: |
RAGD, bA11D8.2.1 |
Location: |
Chr.6:89363509-89412470 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRAGD | XM_005248755.4 | 6 | 5809 | XP_005248812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890901 | Chr.6:89204468 - 90033564 on Build GRCh38 | Gain |
|
| nsv1027007 | Chr.6:89099218 - 89628146 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map