Assay Details
Target Gene Details
Entrez Gene ID: | 167 |
Gene Name: | cysteine rich secretory protein 1 |
Gene Aliases: |
AEGL1, ARP, CRISP-1, HEL-S-57, HSCRISP1D, HSCRISP1G, HUMARP |
Location: |
Chr.6:49834257-49877096 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRISP1 | NM_001131.2 | NP_001122.2 | ||
| NM_001205220.1 | NP_001192149.1 | |||
| NM_170609.1 | NP_733758.1 | |||
| XM_011514315.2 | XP_011512617.1 | |||
| XM_017010320.1 | XP_016865809.1 | |||
| XM_017010321.1 | XP_016865810.1 | |||
| AK292497.1 | ||||
| D38451.1 | BAA07483.1 | |||
| GQ472203.1 | ||||
| S80310.1 | AAB35899.1 | |||
| X95237.1 | CAA64524.1 | |||
| X95238.1 | CAA64525.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608949 | Chr.6:49690413 - 49880115 on Build GRCh38 | Loss |
|
| esv3608951 | Chr.6:49802958 - 49838775 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map