Assay Details
Target Gene Details
Entrez Gene ID: | 10492 |
Gene Name: | synaptotagmin binding cytoplasmic RNA interacting protein |
Gene Aliases: |
GRY-RBP, GRYRBP, HNRNPQ, HNRPQ1, NSAP1, PP68, hnRNP-Q |
Location: |
Chr.6:85607784-85643862 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNCRIP | NM_001159673.1 | 11 | 2304 | NP_001153145.1 |
| NM_001159674.1 | 12 | 2327 | NP_001153146.1 | |
| NM_001159676.1 | 11 | 2335 | NP_001153148.1 | |
| NM_001159677.1 | 12 | 2867 | NP_001153149.1 | |
| NM_001253771.1 | 11 | 2246 | NP_001240700.1 | |
| XM_005248635.4 | 12 | 2485 | XP_005248692.1 | |
| XM_005248636.2 | 10 | 4775 | XP_005248693.1 | |
| XM_005248637.2 | 11 | 4508 | XP_005248694.1 | |
| XM_017010176.1 | 12 | 2630 | XP_016865665.1 | |
| XM_017010177.1 | 12 | 2627 | XP_016865666.1 | |
| XM_017010178.1 | 12 | 2448 | XP_016865667.1 | |
| XM_017010179.1 | 12 | 2420 | XP_016865668.1 | |
| AB209098.1 | 11 | 2244 | BAD92335.1 | |
| BC032643.1 | 12 | 2318 | AAH32643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5383 | Chr.6:85593505 - 85638305 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map