Assay Details
Target Gene Details
Entrez Gene ID: | 60481 |
Gene Name: | ELOVL fatty acid elongase 5 |
Gene Aliases: |
HELO1, SCA38, dJ483K16.1 |
Location: |
Chr.6:53267398-53349179 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL5 | NM_001242828.1 | NP_001229757.1 | ||
| NM_001242830.1 | NP_001229759.1 | |||
| NM_001301856.1 | NP_001288785.1 | |||
| NM_021814.4 | NP_068586.1 | |||
| AB209798.1 | BAD93035.1 | |||
| AF231981.1 | AAF70631.1 | |||
| AF338241.1 | AAM00193.1 | |||
| AK074889.1 | BAC11270.1 | |||
| AK125098.1 | ||||
| AK299674.1 | ||||
| AK302948.1 | ||||
| AL136939.1 | CAB66873.1 | |||
| BC067123.1 | AAH67123.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033994 | Chr.6:53225739 - 53458464 on Build GRCh38 | Gain |
|
| esv2763940 | Chr.6:53228517 - 53622284 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map