Assay Details
Target Gene Details
Entrez Gene ID: | 8737 |
Gene Name: | receptor interacting serine/threonine kinase 1 |
Gene Aliases: |
RIP, RIP-1, RIP1 |
Location: |
Chr.6:3063888-3115187 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIPK1 | NM_001317061.1 | NP_001303990.1 | ||
| NM_003804.4 | NP_003795.2 | |||
| XM_005249457.3 | XP_005249514.2 | |||
| XM_006715237.3 | XP_006715300.2 | |||
| XM_017011405.1 | XP_016866894.1 | |||
| AB208926.1 | BAD92163.1 | |||
| AK304701.1 | ||||
| AK314176.1 | ||||
| BC126254.1 | ||||
| BC126256.1 | ||||
| U50062.1 | AAC32232.1 |
More Information
Set Membership: |
|

Genomic Map