Assay Details
Target Gene Details
Entrez Gene ID: | 105375787 |
Gene Name: | uncharacterized LOC105375787 |
Gene Aliases: |
- |
Location: |
Chr.8:141254567-141256817 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375787 | NM_001317971.1 | NP_001304900.1 | ||
| BF510726.1 |
Target Gene Details
Entrez Gene ID: | 57210 |
Gene Name: | solute carrier family 45 member 4 |
Gene Aliases: |
- |
Location: |
Chr.8:141207166-141308321 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC45A4 | XM_011517169.2 | XP_011515471.1 | ||
| XM_011517170.2 | XP_011515472.1 | |||
| XM_011517171.2 | XP_011515473.1 | |||
| XM_011517172.2 | XP_011515474.1 | |||
| XM_011517173.2 | XP_011515475.1 | |||
| XM_011517176.2 | XP_011515478.1 | |||
| XM_011517179.2 | XP_011515481.1 | |||
| XM_017013648.1 | XP_016869137.1 | |||
| XM_017013649.1 | XP_016869138.1 | |||
| XM_017013650.1 | XP_016869139.1 | |||
| CX166285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522318 | Chr.8:141243157 - 141291925 on Build GRCh38 | Loss |
|
| esv2737756 | Chr.8:140419230 - 141917755 on Build GRCh38 | Deletion |
|
| nsv612568 | Chr.8:140534649 - 141326581 on Build GRCh38 | Gain |
|
| nsv831483 | Chr.8:141123462 - 141290370 on Build GRCh38 | Gain |
|
| nsv1126125 | Chr.8:140419236 - 141917668 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map