Assay Details
Target Gene Details
Entrez Gene ID: | 65986 |
Gene Name: | zinc finger and BTB domain containing 10 |
Gene Aliases: |
RINZF |
Location: |
Chr.8:80485619-80526265 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB10 | NM_001277145.1 | NP_001264074.1 | ||
| AK056913.1 | BAB71309.1 |
More Information
Set Membership: |
|

Genomic Map