Assay Details
Target Gene Details
Entrez Gene ID: | 90459 |
Gene Name: | exoribonuclease 1 |
Gene Aliases: |
3'HEXO, HEXO, THEX1 |
Location: |
Chr.8:8892225-9100262 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERI1 | XM_017013949.1 | XP_016869438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv972228 | Chr.8:8895187 - 8901517 on Build GRCh38 | Deletion |
|
| nsv610190 | Chr.8:8886730 - 8909371 on Build GRCh38 | Loss |
|
| nsv831228 | Chr.8:8750877 - 8933280 on Build GRCh38 | Loss |
|
| esv3616156 | Chr.8:8900075 - 8906494 on Build GRCh38 | Loss |
|
| nsv831229 | Chr.8:8894013 - 9068725 on Build GRCh38 | Gain+Loss |
|
| nsv1018828 | Chr.8:8752370 - 8933150 on Build GRCh38 | Loss |
|
| esv3616155 | Chr.8:8898539 - 8904125 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map