Assay Details
Target Gene Details
Entrez Gene ID: | 51305 |
Gene Name: | potassium two pore domain channel subfamily K member 9 |
Gene Aliases: |
K2p9.1, KT3.2, TASK-3, TASK3 |
Location: |
Chr.8:139600838-139703135 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK9 | NR_104210.1 | |||
| XM_011517101.2 | XP_011515403.1 | |||
| XM_011517102.2 | XP_011515404.1 | |||
| XM_011517103.2 | XP_011515405.1 | |||
| XM_017013530.1 | XP_016869019.1 | |||
| XM_017013531.1 | XP_016869020.1 | |||
| AF257080.1 | AAG33126.1 | |||
| AK090707.1 | ||||
| BC075079.2 | AAH75079.1 | |||
| BC075080.2 | AAH75080.1 | |||
| BC112063.1 | AAI12064.1 | |||
| BC112065.1 | AAI12066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6423 | Chr.8:139573049 - 139617462 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map