Assay Details
Target Gene Details
Entrez Gene ID: | 55638 |
Gene Name: | syntabulin |
Gene Aliases: |
GOLSYN, OCSYN, SNPHL |
Location: |
Chr.8:109574176-109691791 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
More Information
Set Membership: |
|

Genomic Map