Assay Details
Target Gene Details
Entrez Gene ID: | 8840 |
Gene Name: | WNT1 inducible signaling pathway protein 1 |
Gene Aliases: |
CCN4, WISP1c, WISP1i, WISP1tc |
Location: |
Chr.8:133191039-133231690 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WISP1 | NM_001204869.1 | NP_001191798.1 | ||
| NM_001204870.1 | NP_001191799.1 | |||
| NM_003882.3 | NP_003873.1 | |||
| NM_080838.2 | NP_543028.1 | |||
| NR_037944.1 | ||||
| AB034725.1 | BAB17849.1 | |||
| AF100779.1 | AAC96321.1 | |||
| AK293031.1 | ||||
| AK301508.1 | ||||
| AK315822.1 | ||||
| AY196486.1 | AAP43924.1 | |||
| AY196487.1 | AAP43925.1 | |||
| AY196488.1 | AAP43926.1 | |||
| BC074840.2 | AAH74840.1 | |||
| BC074841.2 | AAH74841.1 | |||
| BX476382.1 | ||||
| DN997366.1 |
More Information
Set Membership: |
|

Genomic Map