Assay Details
Target Gene Details
Entrez Gene ID: | 83481 |
Gene Name: | epiplakin 1 |
Gene Aliases: |
EPIPL, EPIPL1 |
Location: |
Chr.8:143857319-143879043 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPPK1 | NM_031308.3 | NP_112598.3 | ||
| XM_017013889.1 | XP_016869378.1 | |||
| XM_017013890.1 | XP_016869379.1 | |||
| XM_017013891.1 | XP_016869380.1 | |||
| XM_017013892.1 | XP_016869381.1 | |||
| BP313273.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3619119 | Chr.8:143874609 - 143883580 on Build GRCh38 | Loss |
|
| dgv12431n54 | Chr.8:143845100 - 144161320 on Build GRCh38 | Loss |
|
| nsv950015 | Chr.8:143866633 - 144125597 on Build GRCh38 | Deletion |
|
| nsv1161815 | Chr.8:143775169 - 144080674 on Build GRCh38 | Deletion |
|
| nsv466009 | Chr.8:143812326 - 143900795 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map