Assay Details
Target Gene Details
Entrez Gene ID: | 54704 |
Gene Name: | pyruvate dehyrogenase phosphatase catalytic subunit 1 |
Gene Aliases: |
PDH, PDP, PDPC, PPM2A, PPM2C |
Location: |
Chr.8:93916855-93926068 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDP1 | NM_001161779.1 | NP_001155251.1 | ||
| NM_001161780.1 | NP_001155252.1 | |||
| NM_001161781.1 | NP_001155253.1 | |||
| NM_018444.3 | NP_060914.2 | |||
| XM_011517135.2 | XP_011515437.1 | |||
| XM_011517136.1 | XP_011515438.1 | |||
| XM_017013587.1 | XP_016869076.1 | |||
| XM_017013588.1 | XP_016869077.1 | |||
| AF155661.1 | AAF67480.1 | |||
| AK057079.1 | ||||
| AK302651.1 | ||||
| BC047619.1 | AAH47619.1 | |||
| BC064978.1 | AAH64978.1 | |||
| DA240240.1 | ||||
| DA507397.1 | ||||
| DA769567.1 | ||||
| DB084539.1 | ||||
| DC369809.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1022473 | Chr.8:91854892 - 94250166 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116385467] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map