Assay Details
Target Gene Details
Entrez Gene ID: | 81790 |
Gene Name: | ring finger protein 170 |
Gene Aliases: |
ADSA, SNAX1 |
Location: |
Chr.8:42849637-42897288 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF170 | NM_001160223.1 | 7 | 3234 | NP_001153695.1 |
| NM_001160224.1 | NP_001153696.1 | |||
| NM_001160225.1 | 7 | 2883 | NP_001153697.1 | |
| NM_030954.3 | 7 | 3004 | NP_112216.3 | |
| NR_027668.1 | 6 | 3160 | ||
| NR_027669.1 | 6 | 2930 | ||
| XM_006716404.2 | 8 | 2991 | XP_006716467.1 | |
| XM_006716405.2 | 5 | 2750 | XP_006716468.1 | |
| XM_011544666.2 | 7 | 2788 | XP_011542968.1 | |
| XM_017013881.1 | 8 | 2931 | XP_016869370.1 | |
| XM_017013882.1 | 6 | 2776 | XP_016869371.1 | |
| AK095625.1 | 1 | 2432 | ||
| BC032393.1 | AAH32393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483102 | Chr.8:42722031 - 42876093 on Build GRCh38 | Loss |
|
| nsv6171 | Chr.8:42853461 - 42887332 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map