Assay Details
Target Gene Details
Entrez Gene ID: | 79776 |
Gene Name: | zinc finger homeobox 4 |
Gene Aliases: |
ZFH4, ZHF4 |
Location: |
Chr.8:76681219-76867285 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFHX4 | NM_024721.4 | NP_078997.4 | ||
| XM_011517592.2 | XP_011515894.1 | |||
| XM_011517593.2 | XP_011515895.1 | |||
| XM_011517594.2 | XP_011515896.1 | |||
| XM_011517595.2 | XP_011515897.1 | |||
| XM_011517596.2 | XP_011515898.1 | |||
| XM_011517597.2 | XP_011515899.1 | |||
| XM_017013845.1 | XP_016869334.1 | |||
| AB083343.2 | ||||
| AK131408.1 | ||||
| DA658176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891416 | Chr.8:75564161 - 76722558 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78023929] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map