Assay Details
Target Gene Details
Entrez Gene ID: | 284 |
Gene Name: | angiopoietin 1 |
Gene Aliases: |
AGP1, AGPT, ANG1 |
Location: |
Chr.8:107249482-107498055 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPT1 | NM_001146.4 | NP_001137.2 | ||
| NM_001199859.2 | NP_001186788.1 | |||
| NM_001314051.1 | NP_001300980.1 | |||
| AB084454.1 | BAB91325.1 | |||
| AK300319.1 | ||||
| AK304717.1 | ||||
| AY055342.1 | AAL59669.1 | |||
| AY121504.1 | AAM81745.1 | |||
| AY124380.1 | AAM92271.1 | |||
| BC152411.1 | ||||
| BC152419.1 | ||||
| BX648814.1 | CAI45984.1 | |||
| D13628.2 | ||||
| U83508.1 | AAB50557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2764100 | Chr.8:107208443 - 107457005 on Build GRCh38 | Gain |
|
| nsv611851 | Chr.8:107212459 - 107473684 on Build GRCh38 | Gain |
|
| nsv1028726 | Chr.8:106804613 - 107380319 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map