Assay Details
Target Gene Details
Entrez Gene ID: | 84750 |
Gene Name: | fucosyltransferase 10 |
Gene Aliases: |
FUCTX |
Location: |
Chr.8:33308102-33473492 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FUT10 | XM_011544676.2 | XP_011542978.1 | ||
| XM_011544677.2 | XP_011542979.1 | |||
| XM_011544678.2 | XP_011542980.1 | |||
| XM_011544681.2 | XP_011542983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6145 | Chr.8:33324644 - 33369268 on Build GRCh38 | Deletion |
|
| esv3576539 | Chr.8:32789248 - 33495217 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72644605] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map