Assay Details
Target Gene Details
Entrez Gene ID: | 81790 |
Gene Name: | ring finger protein 170 |
Gene Aliases: |
ADSA, SNAX1 |
Location: |
Chr.8:42849637-42897288 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF170 | NM_001160224.1 | NP_001153696.1 | ||
| BC032393.1 | AAH32393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483102 | Chr.8:42722031 - 42876093 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map