Assay Details
Target Gene Details
Entrez Gene ID: | 51571 |
Gene Name: | family with sequence similarity 49 member B |
Gene Aliases: |
BM-009, L1 |
Location: |
Chr.8:129839593-130016697 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM49B | NM_001256763.1 | 13 | 3681 | NP_001243692.1 |
| NM_016623.4 | 12 | 3594 | NP_057707.3 | |
| NR_046359.1 | 15 | 3947 | ||
| NR_046360.1 | 15 | 3857 | ||
| NR_046361.1 | 14 | 3847 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576607 | Chr.8:129779957 - 129839982 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115949023] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map