Assay Details
Target Gene Details
Entrez Gene ID: | 84376 |
Gene Name: | hook microtubule-tethering protein 3 |
Gene Aliases: |
HK3 |
Location: |
Chr.8:42896890-43030539 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOOK3 | NM_032410.3 | NP_115786.1 | ||
| AK309224.1 | ||||
| BC048304.1 | AAH48304.1 | |||
| BC056146.1 | AAH56146.1 |
More Information
Set Membership: |
|

Genomic Map