Assay Details
Target Gene Details
Entrez Gene ID: | 157556 |
Gene Name: | BAALC antisense RNA 2 |
Gene Aliases: |
BAALCOS, C8orf56 |
Location: |
Chr.8:103132963-103141342 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAALC-AS2 | NR_027071.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831413 | Chr.8:103072913 - 103257561 on Build GRCh38 | Gain |
|

Genomic Map